Tisk…

4-Chloro-2-fluoro-5-methylbenzoyl chloride

Dodavatel: Apollo Scientific

PC303326-5G
APOSPC303326-5GEA 19580 CZK
APOSPC303326-5G
4-Chloro-2-fluoro-5-methylbenzoyl chloride
4-Chloro-2-fluoro-5-methylbenzoyl chloride
Vzorec: C₈H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD13174586
CAS číslo: 204778-68-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR