Tisk…

4-Fluoro-3-methyl-2-(trifluoromethyl)phenylacetic acid 97%

Dodavatel: Apollo Scientific

PC303224-1G PC303224-250MG
APOSPC303224-1GEA 25590 CZK
APOSPC303224-1G APOSPC303224-250MG
4-Fluoro-3-methyl-2-(trifluoromethyl)phenylacetic acid 97%
4-Fluoro-3-methyl-2-(trifluoromethyl)phenylacetic acid
Vzorec: C₁₀H₈F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD28054204
CAS číslo: 1706439-04-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR