Tisk…

3,4-Dichloro-5-(trifluoromethoxy)benzoyl chloride 97%

Dodavatel: Apollo Scientific

PC303171-1G PC303171-250MG
APOSPC303171-1GEA 17660 CZK
APOSPC303171-1G APOSPC303171-250MG
3,4-Dichloro-5-(trifluoromethoxy)benzoyl chloride 97%
3,4-Dichloro-5-(trifluoromethoxy)benzoyl chloride
Vzorec: C₈H₂Cl₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28054195
CAS číslo: 1706458-00-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR