Tisk…

2-Fluoro-5-methyl-3-(trifluoromethyl)benzyl bromide 97%

Dodavatel: Apollo Scientific

PC303170-1G PC303170-250MG
APOSPC303170-1GEA 17660 CZK
APOSPC303170-1G APOSPC303170-250MG
2-Fluoro-5-methyl-3-(trifluoromethyl)benzyl bromide 97%
2-Fluoro-5-methyl-3-(trifluoromethyl)benzyl bromide
Vzorec: C₉H₇BrF₄
Storage Temperature: Ambient
MDL Number: MFCD28054188
CAS číslo: 1706438-74-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR