Tisk…

4-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid 97%

Dodavatel: Apollo Scientific

PC303137-1G PC303137-5G
APOSPC303137-1GEA 8830 CZK
APOSPC303137-1G APOSPC303137-5G
4-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid 97%
4-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid
Vzorec: C₉H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD27986860
CAS číslo: 1706436-25-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR