Tisk…

4-n-Butoxy-2,6-difluorobenzoic acid 98%

Dodavatel: Apollo Scientific

PC303075-10G PC303075-5G PC303075-1G
APOSPC303075-10GEA 9330 CZK
APOSPC303075-10G APOSPC303075-5G APOSPC303075-1G
4-n-Butoxy-2,6-difluorobenzoic acid 98%
4-n-Butoxy-2,6-difluorobenzoic acid
Vzorec: C₁₁H₁₂F₂O₃
Bod tání: 112…115 °C
Storage Temperature: Ambient
MDL Number: MFCD22201018
CAS číslo: 123843-56-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR