Tisk…

4-Fluoro-2-(trifluoromethoxy)benzonitrile 97%

Dodavatel: Apollo Scientific

PC302989-5G PC302989-1G PC302989-250MG
APOSPC302989-5GEA 18090 CZK
APOSPC302989-5G APOSPC302989-1G APOSPC302989-250MG
4-Fluoro-2-(trifluoromethoxy)benzonitrile 97%
4-Fluoro-2-(trifluoromethoxy)benzonitrile
Vzorec: C₈H₃F₄NO
Storage Temperature: Ambient
MDL Number: MFCD19687191
CAS číslo: 1323966-32-0
UN: 3276
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR