Tisk…

4-(Difluoromethoxy)-3-methoxybenzoic acid 98%

Dodavatel: Apollo Scientific

PC302966-5G PC302966-1G
APOSPC302966-5GEA 5060 CZK
APOSPC302966-5G APOSPC302966-1G
4-(Difluoromethoxy)-3-methoxybenzoic acid 98%
4-(Difluoromethoxy)-3-methoxybenzoic acid
Vzorec: C₉H₈F₂O₄
MW: 218.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03964657
CAS číslo: 162401-59-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR