Tisk…

3-(2,3,5-Trifluorophenyl)propionic acid 98%

Dodavatel: Apollo Scientific

PC302936-10G PC302936-5G PC302936-1G
APOSPC302936-10GEA 9000 CZK
APOSPC302936-10G APOSPC302936-5G APOSPC302936-1G
3-(2,3,5-Trifluorophenyl)propionic acid 98%
3-(2,3,5-Trifluorophenyl)propionic acid
Vzorec: C₉H₇F₃O₂
MW: 204.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660215
CAS číslo: 886499-45-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR