Tisk…

2-Chloro-4-(trifluoromethyl)benzyl alcohol 98%

Dodavatel: Apollo Scientific

Synonyms: 2-Chloro-4-(trifluoromethyl)benzylalcohol

PC302909-1G
APOSPC302909-1GEA 787 CZK
APOSPC302909-1G
2-Chloro-4-(trifluoromethyl)benzyl alcohol 98%
2-Chloro-4-(trifluoromethyl)benzyl alcohol
Vzorec: C₈H₆ClF₃O
MW: 210.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972757
CAS číslo: 56456-51-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR