Tisk…

3-Chloro-2-fluoro-4-(trifluoromethyl)benzoic acid

Dodavatel: Apollo Scientific
Varování

PC302830-10G PC302830-1G PC302830-5G
APOSPC302830-10GEA 28110 CZK
APOSPC302830-10G APOSPC302830-1G APOSPC302830-5G
3-Chloro-2-fluoro-4-(trifluoromethyl)benzoic acid
3-Chloro-2-fluoro-4-(trifluoromethyl)benzoic acid
Vzorec: C₈H₃ClF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD23795337
CAS číslo: 1431329-63-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR