Tisk…

3-Chloro-4-ethoxy-5-fluorobenzyl alcohol 97%

Dodavatel: Apollo Scientific

PC302771-10G PC302771-5G
APOSPC302771-10GEA 6850 CZK
APOSPC302771-10G APOSPC302771-5G
3-Chloro-4-ethoxy-5-fluorobenzyl alcohol 97%
3-Chloro-4-ethoxy-5-fluorobenzyl alcohol
Vzorec: C₉H₁₀ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD09832341
CAS číslo: 1017778-88-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR