Tisk…

5-Fluoro-2-(trifluoromethoxy)aniline 98%

Dodavatel: Apollo Scientific

PC302761-1G PC302761-5G PC302761-10G
APOSPC302761-1GEA 4200 CZK
APOSPC302761-1G APOSPC302761-5G APOSPC302761-10G
5-Fluoro-2-(trifluoromethoxy)aniline 98%
5-Fluoro-2-(trifluoromethoxy)aniline
Vzorec: C₇H₅F₄NO
Storage Temperature: Ambient
MDL Number: MFCD16652514
CAS číslo: 123572-63-4
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR