Tisk…

(4-Methyl-3-(trifluoromethyl)phenyl)acetic acid 98%

Dodavatel: Apollo Scientific
Varování

PC302757-1G PC302757-5G PC302757-10G
APOSPC302757-1GEA 2610 CZK
APOSPC302757-1G APOSPC302757-5G APOSPC302757-10G
(4-Methyl-3-(trifluoromethyl)phenyl)acetic acid 98%
(4-Methyl-3-(trifluoromethyl)phenyl)acetic acid
Vzorec: C₁₀H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09832293
CAS číslo: 1000544-72-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR