Tisk…

4-Methoxy-2-(trifluoromethyl)benzyl alcohol 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Methoxy-2-(trifluoromethyl)phenyl)methanol

PC302755-10G PC302755-5G PC302755-1G
APOSPC302755-10GEA 7080 CZK
APOSPC302755-10G APOSPC302755-5G APOSPC302755-1G
4-Methoxy-2-(trifluoromethyl)benzyl alcohol 98%
4-Methoxy-2-(trifluoromethyl)benzyl alcohol
Vzorec: C₉H₉F₃O₂
MW: 206.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06202983
CAS číslo: 773871-39-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR