Tisk…

2,4-Difluoro-3-methoxybenzoyl chloride 98%

Dodavatel: Apollo Scientific

PC302662-25G PC302662-5G PC302662-1G
APOSPC302662-25GEA 38350 CZK
APOSPC302662-25G APOSPC302662-5G APOSPC302662-1G
2,4-Difluoro-3-methoxybenzoyl chloride 98%
2,4-Difluoro-3-methoxybenzoyl chloride
Vzorec: C₈H₅ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04115933
CAS číslo: 221221-11-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR