Tisk…

2-Methoxy-4-(trifluoromethyl)benzyl alcohol 98%

Dodavatel: Apollo Scientific
Varování

PC302642-5G PC302642-1G
APOSPC302642-5GEA 5060 CZK
APOSPC302642-5G APOSPC302642-1G
2-Methoxy-4-(trifluoromethyl)benzyl alcohol 98%
2-Methoxy-4-(trifluoromethyl)benzyl alcohol
Vzorec: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04115979
CAS číslo: 286441-68-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR