Tisk…

3-Fluoro-4-(trifluoromethoxy)benzamide 98%

Dodavatel: Apollo Scientific

PC302550-1G PC302550-5G
APOSPC302550-1GEA 3310 CZK
APOSPC302550-1G APOSPC302550-5G
3-Fluoro-4-(trifluoromethoxy)benzamide 98%
3-Fluoro-4-(trifluoromethoxy)benzamide
Vzorec: C₈H₅F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD06660208
CAS číslo: 886499-18-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR