Tisk…

2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid 98%

Dodavatel: Apollo Scientific
Varování

PC302458-5G PC302458-1G
APOSPC302458-5GEA 3310 CZK
APOSPC302458-5G APOSPC302458-1G
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid 98%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid
Vzorec: C₉H₆F₄O₃
MW: 238.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201034
CAS číslo: 1373920-90-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR