Tisk…

3-Fluoro-5-(trifluoromethoxy)benzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC302305-1G PC302305-5G
APOSPC302305-1GEA 11150 CZK
APOSPC302305-1G APOSPC302305-5G
3-Fluoro-5-(trifluoromethoxy)benzylamine 98%
3-Fluoro-5-(trifluoromethoxy)benzylamine
Vzorec: C₈H₇F₄NO
Storage Temperature: Ambient
MDL Number: MFCD18783229
CAS číslo: 1353000-19-7
UN: 3259
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR