Tisk…

3,5-Difluoro-o-toluic acid 98%

Dodavatel: Apollo Scientific
Varování

PC302272-1G PC302272-5G PC302272-10G
APOSPC302272-1GEA 3640 CZK
APOSPC302272-1G APOSPC302272-5G APOSPC302272-10G
3,5-Difluoro-o-toluic acid 98%
3,5-Difluoro-o-toluic acid
Vzorec: C₈H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09832334
CAS číslo: 1003710-06-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR