Tisk…

2-Methoxy-6-(trifluoromethyl)benzyl bromide 98%

Dodavatel: Apollo Scientific

PC302259-1G PC302259-5G PC302259-10G
APOSPC302259-1GEA 4200 CZK
APOSPC302259-1G APOSPC302259-5G APOSPC302259-10G
2-Methoxy-6-(trifluoromethyl)benzyl bromide 98%
2-Methoxy-6-(trifluoromethyl)benzyl bromide
Vzorec: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09832326
CAS číslo: 1017779-04-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR