Tisk…

3-Chloro-5-(pentafluorothio)benzoyl chloride 97%

Dodavatel: Apollo Scientific

PC302196-1G
APOSPC302196-1GEA 12010 CZK
APOSPC302196-1G
3-Chloro-5-(pentafluorothio)benzoyl chloride 97%
3-Chloro-5-(pentafluorothio)benzoyl chloride
Vzorec: C₇H₃Cl₂F₅OS
Storage Temperature: Ambient
MDL Number: MFCD16652451
CAS číslo: 1240257-81-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR