Tisk…

2-Chloro-4,5-difluoro-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302126-1G PC302126-5G PC302126-10G
APOSPC302126-1GEA 3310 CZK
APOSPC302126-1G APOSPC302126-5G APOSPC302126-10G
2-Chloro-4,5-difluoro-DL-phenylalanine
2-Chloro-4,5-difluoro-DL-phenylalanine
Vzorec: C₉H₈ClF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD06660323
CAS číslo: 1259983-39-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR