Tisk…

(3-Chloro-2,4-difluorophenyl)acetic acid 97%

Dodavatel: Apollo Scientific

PC302087-1G PC302087-5G PC302087-250MG
APOSPC302087-1GEA 2480 CZK
APOSPC302087-1G APOSPC302087-5G APOSPC302087-250MG
(3-Chloro-2,4-difluorophenyl)acetic acid 97%
(3-Chloro-2,4-difluorophenyl)acetic acid
Vzorec: C₈H₅ClF₂O₂
Bod tání: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD06660337
CAS číslo: 886761-66-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR