Tisk…

2-Methoxy-3-(trifluoromethyl)benzyl bromide 98%

Dodavatel: Apollo Scientific

PC302078-1G PC302078-5G PC302078-25G PC302078-100G
APOSPC302078-1GEA 1990 CZK
APOSPC302078-1G APOSPC302078-5G APOSPC302078-25G APOSPC302078-100G
2-Methoxy-3-(trifluoromethyl)benzyl bromide 98%
2-Methoxy-3-(trifluoromethyl)benzyl bromide
Vzorec: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09832315
CAS číslo: 1017778-77-6
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR