Tisk…

3,4-Difluorobenzamidoxime

Dodavatel: Apollo Scientific

PC302036-10G PC302036-50G
APOSPC302036-10GEA 2280 CZK
APOSPC302036-10G APOSPC302036-50G
3,4-Difluorobenzamidoxime
3,4-Difluorobenzamidoxime
Vzorec: C₇H₆F₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD06660194
CAS číslo: 500024-74-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR