Tisk…

2-Chloro-6-(trifluoromethyl)phenylacetonitrile 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC302025-1G PC302025-5G PC302025-25G PC302025-100G
APOSPC302025-1GEA 3640 CZK
APOSPC302025-1G APOSPC302025-5G APOSPC302025-25G APOSPC302025-100G
2-Chloro-6-(trifluoromethyl)phenylacetonitrile 97%
2-Chloro-6-(trifluoromethyl)phenylacetonitrile
Vzorec: C₉H₅ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD06660233
CAS číslo: 633317-69-8
UN: 3439
ADR: 6.1,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR