Tisk…

Ethyl 2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate

Dodavatel: Apollo Scientific
Varování

PC301029-250MG PC301029-1G
APOSPC301029-250MGEA 3940 CZK
APOSPC301029-250MG APOSPC301029-1G
Ethyl 2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Vzorec: C₁₁H₁₀F₂N₂O₂
MDL Number: MFCD32689358
CAS číslo: 2451256-48-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR