Tisk…

(4-Bromo-2-fluoro-5-methylphenoxy)acetonitrile

Dodavatel: Apollo Scientific

PC301020-1G
APOSPC301020-1GEA 10550 CZK
APOSPC301020-1G
(4-Bromo-2-fluoro-5-methylphenoxy)acetonitrile
(4-Bromo-2-fluoro-5-methylphenoxy)acetonitrile
Vzorec: C₉H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD31725301
CAS číslo: 2270913-02-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR