Tisk…

1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanol

Dodavatel: Apollo Scientific
Varování

PC300994-1G
APOSPC300994-1GEA 4630 CZK
APOSPC300994-1G
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanol
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanol
Vzorec: C₁₄H₁₃FO
MW: 216,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18089274
CAS číslo: 56430-67-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR