Tisk…

2-Chloro-N-(3-fluoro-4-methylphenyl)acetamide

Dodavatel: Apollo Scientific

PC300959-5G
APOSPC300959-5GEA 2200 CZK
APOSPC300959-5G
2-Chloro-N-(3-fluoro-4-methylphenyl)acetamide
2-Chloro-N-(3-fluoro-4-methylphenyl)acetamide
Vzorec: C₉H₉ClFNO
Storage Temperature: Ambient
MDL Number: MFCD03152474
CAS číslo: 100599-62-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR