Tisk…

Methyl 2,5-difluoro-4-(4-methylpiperazin-1-yl)benzoate

Dodavatel: Apollo Scientific

PC300788-1G PC300788-5G PC300788-250MG
APOSPC300788-1GEA 8270 CZK
APOSPC300788-1G APOSPC300788-5G APOSPC300788-250MG
Methyl 2,5-difluoro-4-(4-methylpiperazin-1-yl)benzoate
Methyl 2,5-difluoro-4-(4-methylpiperazin-1-yl)benzoate
Vzorec: C₁₃H₁₆F₂N₂O₂
MW: 270.275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103349
CAS číslo: 1858251-84-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR