Tisk…

Methyl 2,5-difluoro-4-methoxybenzoate

Dodavatel: Apollo Scientific

PC300785-1G PC300785-5G PC300785-250MG
APOSPC300785-1GEA 8170 CZK
APOSPC300785-1G APOSPC300785-5G APOSPC300785-250MG
Methyl 2,5-difluoro-4-methoxybenzoate
Methyl 2,5-difluoro-4-methoxybenzoate
Vzorec: C₉H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD18399129
CAS číslo: 1261830-21-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR