Tisk…

8-Chloro-3-nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine

Dodavatel: Apollo Scientific
Varování

PC300756-1G
APOSPC300756-1GEA 7340 CZK
APOSPC300756-1G
8-Chloro-3-nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
8-Chloro-3-nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Vzorec: C₈H₃ClF₃N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD27995619
CAS číslo: 1858241-44-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR