Tisk…

2-[(2,4-Difluorophenoxy)methyl]oxirane

Dodavatel: Apollo Scientific

PC300681-1G PC300681-5G PC300681-10G
APOSPC300681-1GEA 4930 CZK
APOSPC300681-1G APOSPC300681-5G APOSPC300681-10G
2-[(2,4-Difluorophenoxy)methyl]oxirane
2-[(2,4-Difluorophenoxy)methyl]oxirane
Vzorec: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09741082
CAS číslo: 169286-61-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR