Tisk…

2-[4-(2-Chloro-6-fluorobenzyl)piperazin-1-yl]ethanol

Dodavatel: Apollo Scientific

PC300651-1G PC300651-10G PC300651-5G
APOSPC300651-1GEA 5630 CZK
APOSPC300651-1G APOSPC300651-10G APOSPC300651-5G
2-[4-(2-Chloro-6-fluorobenzyl)piperazin-1-yl]ethanol
2-[4-(2-Chloro-6-fluorobenzyl)piperazin-1-yl]ethanol
Vzorec: C₁₃H₁₈ClFN₂O
Storage Temperature: Ambient
MDL Number: MFCD01453128
CAS číslo: 416894-47-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR