Tisk…

Ethyl 3,8-dichloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

Dodavatel: Apollo Scientific
Varování

PC300582-1G
APOSPC300582-1GEA 1660 CZK
APOSPC300582-1G
Ethyl 3,8-dichloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3,8-dichloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Vzorec: C₁₁H₇Cl₂F₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD17168289
CAS číslo: 1355171-49-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR