Tisk…

[2-Morpholin-4-yl-5-(trifluoromethyl)phenyl]methanol

Dodavatel: Apollo Scientific
Varování

PC300518-1G PC300518-250MG
APOSPC300518-1GEA 2900 CZK
APOSPC300518-1G APOSPC300518-250MG
[2-Morpholin-4-yl-5-(trifluoromethyl)phenyl]methanol
[2-Morpholin-4-yl-5-(trifluoromethyl)phenyl]methanol
Vzorec: C₁₂H₁₄F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD09025885
CAS číslo: 886851-51-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR