Tisk…

2-Chloro-4-(trifluoromethyl)phenylhydrazine hydrochloride 97%

Dodavatel: Apollo Scientific

PC2897-5G
APOSPC2897-5GEA 5000 CZK
APOSPC2897-5G
2-Chloro-4-(trifluoromethyl)phenylhydrazine hydrochloride 97%
2-Chloro-4-(trifluoromethyl)phenylhydrazine hydrochloride
Vzorec: C₇H₆ClF₃N₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD03094209
CAS číslo: 1029649-46-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR