Tisk…

2,4-Difluorophenyl isocyanate 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC2875H-100G PC2875H-25G PC2875H-5G
APOSPC2875H-100GEA 6620 CZK
APOSPC2875H-100G APOSPC2875H-25G APOSPC2875H-5G
2,4-Difluorophenyl isocyanate 98%
2,4-Difluorophenyl isocyanate

  • May form small amount of sediment.

Vzorec: C₇H₃F₂NO
Bod varu: 42 °C
Hustota: 1,309 g/cm³ (25 °C)
Bod vzplanutí: 55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001997
CAS číslo: 59025-55-7
UN: 2478
ADR: 3+6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR