Tisk…

8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3-amine hydrobromide

Dodavatel: Apollo Scientific
Varování

PC28284-1G PC28284-250MG
APOSPC28284-1GEA 2980 CZK
APOSPC28284-1G APOSPC28284-250MG
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3-amine hydrobromide
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3-amine hydrobromide
Vzorec: C₇H₄ClF₃N₄·BrH
Storage Temperature: Ambient
MDL Number: MFCD12546141
CAS číslo: 2379918-43-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR