Tisk…

2-(4-Fluorophenyl)imidazo[1,2-a]pyridine-8-carboxylic acid, tech

Dodavatel: Apollo Scientific

PC28133-1G
APOSPC28133-1GEA 44640 CZK
APOSPC28133-1G
2-(4-Fluorophenyl)imidazo[1,2-a]pyridine-8-carboxylic acid, tech
2-(4-Fluorophenyl)imidazo[1,2-a]pyridine-8-carboxylic acid
Vzorec: C₁₄H₉FN₂O₂
MW: 256,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05256204
CAS číslo: 133427-95-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR