Tisk…

1-[2-(4-Fluorophenyl)-1,3-thiazol-5-yl]ethan-1-one, tech

Dodavatel: Apollo Scientific

PC28104-1G
APOSPC28104-1GEA 44640 CZK
APOSPC28104-1G
1-[2-(4-Fluorophenyl)-1,3-thiazol-5-yl]ethan-1-one, tech
1-[2-(4-Fluorophenyl)-1,3-thiazol-5-yl]ethan-1-one
Vzorec: C₁₁H₈FNOS
MW: 221,251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16311596
CAS číslo: 1226001-23-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR