Tisk…

2-Amino-6-chloro-4-fluorotoluene 95+%

Dodavatel: Apollo Scientific

PC2789-250MG PC2789-1G PC2789-5G
APOSPC2789-250MGEA 5030 CZK
APOSPC2789-250MG APOSPC2789-1G APOSPC2789-5G
2-Amino-6-chloro-4-fluorotoluene 95+%
2-Amino-6-chloro-4-fluorotoluene
Vzorec: C₇H₇ClFN
Storage Temperature: Ambient
MDL Number: MFCD03094195
CAS číslo: 886761-87-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR