Tisk…

α-Bromo-2,6-difluorotoluene 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC2743-25G PC2743-100G
APOSPC2743-25GEA 1210 CZK
APOSPC2743-25G APOSPC2743-100G
α-Bromo-2,6-difluorotoluene 98%
α-Bromo-2,6-difluorotoluene

Can protect alcohols as 2,6-difluorobenzyl ethers see : Helv. Chim. Acta, 79, 1159 (1996), these ethersunlike benzyl ethers are stable during cationic reactions.

Vzorec: C₇H₅BrF₂
Bod tání: 51…55 °C
Bod vzplanutí: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000329
CAS číslo: 85118-00-9
EINECS: 285-652-6
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR