Tisk…

4-Iodo-β,β,β-trifluorophenetole

Dodavatel: Apollo Scientific
Varování

PC2634-1G PC2634-5G
APOSPC2634-1GEA 3610 CZK
APOSPC2634-1G APOSPC2634-5G
4-Iodo-β,β,β-trifluorophenetole
4-Iodo-β,β,β-trifluorophenetole
Vzorec: C₈H₆F₃IO
Storage Temperature: Ambient
MDL Number: MFCD08458086
CAS číslo: 530080-17-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR