Tisk…

3-Bromo-5-fluorobenzyl alcohol 98%

Dodavatel: Apollo Scientific
Varování

PC2406-1G PC2406-25G PC2406-5G
APOSPC2406-1GEA 622 CZK
APOSPC2406-1G APOSPC2406-25G APOSPC2406-5G
3-Bromo-5-fluorobenzyl alcohol 98%
3-Bromo-5-fluorobenzyl alcohol
Vzorec: C₇H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD04116320
CAS číslo: 216755-56-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR