Tisk…

3,5-Bis(trifluoromethyl)fluorobenzene 98%

Dodavatel: Apollo Scientific
Varování

PC2283-5G PC2283-100G PC2283-25G
APOSPC2283-5GEA 622 CZK
APOSPC2283-5G APOSPC2283-100G APOSPC2283-25G
3,5-Bis(trifluoromethyl)fluorobenzene 98%
3,5-Bis(trifluoromethyl)fluorobenzene
Vzorec: C₈H₃F₇
Storage Temperature: Ambient
MDL Number: MFCD00017997
CAS číslo: 35564-19-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR