Tisk…

N-(4-Fluorophenyl)-3-methyl-[1,2,4]triazolo[4,3-a]pyridine-8-sulphonamide

Dodavatel: Apollo Scientific

PC210050-1G
APOSPC210050-1GEA 22220 CZK
APOSPC210050-1G
N-(4-Fluorophenyl)-3-methyl-[1,2,4]triazolo[4,3-a]pyridine-8-sulphonamide
N-(4-Fluorophenyl)-3-methyl-[1,2,4]triazolo[4,3-a]pyridine-8-sulphonamide
Vzorec: C₁₃H₁₁FN₄O₂S
MW: 306.315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18804108
CAS číslo: 1291487-08-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR